siRNA Details
| Gene Symbol: Mamdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174857.3 | NP_777282.1 | 3 | 504 | ||
| GenBank mRNA | AK004794.1 | - | 3 | 624 | ||
| AK035566.1 | - | 3 | 505 | |||
| AK154219.1 | - | 3 | 581 | |||
| BC042773.1 | - | 3 | 533 | 4019075 | ||

Mamdc2
Genomic Map