siRNA Details
| Gene Symbol: Rgl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023622.4 | NP_076111.2 | 3 | 221 | ||
| GenBank mRNA | AF237669.1 | - | 3 | 180 | ||
| AF239661.1 | - | 3 | 222 | |||
| AK053550.1 | - | 3 | 208 | |||
| AK134654.1 | - | 203 | ||||
| AK165023.1 | - | 3 | 214 | |||
| AK165746.1 | - | 3 | 214 | |||

Rgl3
Genomic Map