siRNA Details
| Gene Symbol: 1300017J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027918.2 | NP_082194.1 | 8 | 1044 | ||
| GenBank mRNA | AK005035.1 | - | 8 | 1045 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1300017J02Rik| Gene Symbol: 1300017J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027918.2 | NP_082194.1 | 8 | 1044 | ||
| GenBank mRNA | AK005035.1 | - | 8 | 1045 | ||