siRNA Details
| Gene Symbol: Cpa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027926.2 | NP_082202.1 | 6, 7 | 608 | ||
| GenBank mRNA | AK030705.1 | - | 6, 7 | 608 | ||
| BC061206.1 | - | 6, 7 | 615 | 30249166 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpa4| Gene Symbol: Cpa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027926.2 | NP_082202.1 | 6, 7 | 608 | ||
| GenBank mRNA | AK030705.1 | - | 6, 7 | 608 | ||
| BC061206.1 | - | 6, 7 | 615 | 30249166 | ||