siRNA Details
Gene Symbol: Cpa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027926.2 | NP_082202.1 | 4 | 357 | ||
GenBank mRNA | AK030705.1 | - | 4 | 357 | ||
BC061206.1 | - | 4 | 364 | 30249166 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cpa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027926.2 | NP_082202.1 | 4 | 357 | ||
GenBank mRNA | AK030705.1 | - | 4 | 357 | ||
BC061206.1 | - | 4 | 364 | 30249166 |