siRNA Details
| Gene Symbol: Ints12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027927.3 | NP_082203.1 | 4 | 415 | ||
| GenBank mRNA | AK003874.1 | - | 4 | 427 | ||
| AK012829.1 | - | 4 | 428 | |||
| AK038713.1 | - | 2 | 3982 | |||
| AK054389.1 | - | 1 | 1288 | |||
| AK155162.1 | - | 4 | 427 | |||
| BC010657.1 | - | 2 | 87 | |||
| BC023801.1 | - | 4 | 415 | 5353058 | ||
| BC046996.1 | - | 4 | 409 | 6494551 | ||

Ints12
Genomic Map