siRNA Details
| Gene Symbol: Ints12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027927.3 | NP_082203.1 | 5, 6 | 690 | ||
| GenBank mRNA | AK003874.1 | - | 5, 6 | 702 | ||
| AK012829.1 | - | 5, 6 | 701 | |||
| AK155162.1 | - | 5, 6 | 702 | |||
| BC010657.1 | - | 3, 4 | 362 | |||
| BC023801.1 | - | 5, 6 | 690 | 5353058 | ||
| BC046996.1 | - | 5, 6 | 684 | 6494551 | ||

Ints12
Genomic Map