siRNA Details
| Gene Symbol: Nup93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172410.2 | NP_765998.1 | 3 | 182 | ||
| GenBank mRNA | AJ505051.1 | - | 2 | 118 | ||
| AK010450.1 | - | 2 | 134 | |||
| AK028864.1 | - | 3 | 183 | |||
| AK129055.1 | - | 2 | 132 | |||
| AK167105.1 | - | 2 | 133 | |||
| AK169693.1 | - | 2 | 152 | |||
| BC023140.1 | - | 2 | 128 | 3991335 | ||

Nup93
Genomic Map