siRNA Details
| Gene Symbol: Nup93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172410.2 | NP_765998.1 | 5, 6 | 467 | ||
| GenBank mRNA | AJ505051.1 | - | 4, 5 | 403 | ||
| AK010450.1 | - | 4, 5 | 418 | |||
| AK028864.1 | - | 5, 6 | 468 | |||
| AK045380.1 | - | 326 | ||||
| AK129055.1 | - | 4, 5 | 417 | |||
| AK167105.1 | - | 4, 5 | 418 | |||
| AK169194.1 | - | 2, 3 | 188 | |||
| AK169693.1 | - | 4, 5 | 437 | |||
| BC023140.1 | - | 4, 5 | 413 | 3991335 | ||

Nup93
Genomic Map