siRNA Details
| Gene Symbol: Kif23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024245.4 | NP_077207.3 | 5 | 575 | ||
| GenBank mRNA | AK013939.1 | - | 5 | 513 | ||
| AK030771.1 | - | 5 | 552 | |||
| BC003363.1 | - | 5 | 548 | |||
| BC047273.1 | - | 5 | 533 | 6440797 | ||

Kif23
Genomic Map