siRNA Details
| Gene Symbol: Fbxo30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168297.1 | NP_001161769.1 | 2 | 326 | ||
| NM_027968.3 | NP_082244.2 | 2 | 360 | |||
| GenBank mRNA | AK006369.1 | - | 2 | 327 | ||
| AK083479.1 | - | 2 | 317 | |||
| AK135274.1 | - | 2 | 319 | |||

Fbxo30
Genomic Map