siRNA Details
| Gene Symbol: 1700023E05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027970.1 | NP_082246.1 | 4 | 760 | ||
| GenBank mRNA | AK006270.1 | - | 4 | 740 | ||
| BC053422.1 | - | 4 | 760 | 6741654 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700023E05Rik| Gene Symbol: 1700023E05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027970.1 | NP_082246.1 | 4 | 760 | ||
| GenBank mRNA | AK006270.1 | - | 4 | 740 | ||
| BC053422.1 | - | 4 | 760 | 6741654 | ||