siRNA Details
| Gene Symbol: Aox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023631.2 | NP_076120.2 | 21 | 2459 | ||
| GenBank mRNA | AF233581.1 | - | 21 | 2386 | ||
| AK158427.1 | - | 21 | 2460 | |||
| AK160953.1 | - | 21 | 2425 | |||
| BC117975.1 | - | 21 | 2386 | 40098789 | ||
| BC117976.1 | - | 21 | 2386 | 40098791 | ||

Aox4
Genomic Map