siRNA Details
| Gene Symbol: 2310003H01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027980.2 | NP_082256.2 | 3 | 809 | ||
| GenBank mRNA | AK009114.1 | - | 3 | 773 | ||
| AK082976.1 | - | 3 | 810 | |||
| AK147900.1 | - | 895 | ||||
| AK148234.1 | - | 3 | 794 | |||
| AK165631.1 | - | 2 | 680 | |||
| BC034203.1 | - | 3 | 680 | 4986832 | ||

2310003H01Rik
Genomic Map