siRNA Details
| Gene Symbol: Apol9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168660.1 | NP_001162131.1 | 3 | 800 | ||
| NM_173743.4 | NP_776104.1 | 3 | 751 | |||
| GenBank mRNA | AK009386.1 | - | 3 | 801 | ||
| AK050167.1 | - | 3 | 753 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apol9b| Gene Symbol: Apol9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168660.1 | NP_001162131.1 | 3 | 800 | ||
| NM_173743.4 | NP_776104.1 | 3 | 751 | |||
| GenBank mRNA | AK009386.1 | - | 3 | 801 | ||
| AK050167.1 | - | 3 | 753 | |||