siRNA Details
| Gene Symbol: Tmem106b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027992.3 | NP_082268.2 | 6 | 845 | ||
| GenBank mRNA | AK009646.1 | - | 6 | 748 | ||
| AK148477.1 | - | 6 | 769 | |||
| BC050246.1 | - | 6 | 762 | 6313498 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem106b| Gene Symbol: Tmem106b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027992.3 | NP_082268.2 | 6 | 845 | ||
| GenBank mRNA | AK009646.1 | - | 6 | 748 | ||
| AK148477.1 | - | 6 | 769 | |||
| BC050246.1 | - | 6 | 762 | 6313498 | ||