siRNA Details
| Gene Symbol: Tmem106b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027992.3 | NP_082268.2 | 3 | 364 | ||
| GenBank mRNA | AI552875.1 | - | 3 | 380 | ||
| AK009646.1 | - | 3 | 267 | |||
| AK018015.1 | - | 3 | 271 | |||
| AK148477.1 | - | 3 | 286 | |||
| BC050246.1 | - | 3 | 281 | 6313498 | ||

Tmem106b
Genomic Map