siRNA Details
| Gene Symbol: Epgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053087.2 | NP_444317.1 | 4 | 317 | ||
| GenBank mRNA | AJ291391.1 | - | 4 | 317 | ||
| AK010136.1 | - | 3 | 265 | |||
| BC119291.1 | - | 4 | 370 | 8734040 | ||
| BC119293.1 | - | 4 | 370 | 8734042 | ||
| BC145041.1 | - | 4 | 417 | |||

Epgn
Genomic Map