siRNA Details
| Gene Symbol: 2310067B10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028014.3 | NP_082290.2 | 15 | 1942 | ||
| GenBank mRNA | AK047622.1 | - | 1 | 76 | ||
| AK155233.1 | - | 15 | 1817 | |||
| BC007157.1 | - | 2 | 188 | |||
| BC053088.1 | - | 15 | 1857 | 5709104 | ||

2310067B10Rik
Genomic Map