siRNA Details
| Gene Symbol: Rnf135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028019.3 | NP_082295.1 | 5 | 1031 | ||
| GenBank mRNA | AK010429.1 | - | 5 | 1033 | ||
| AK150921.1 | - | 5 | 1032 | |||
| AK152096.1 | - | 5 | 1024 | |||
| BC025209.1 | - | 1 | 194 | |||
| BC096385.1 | - | 5 | 1025 | 30744704 | ||
| BC132307.1 | - | 5 | 1027 | 40130584 | ||
| BC138303.1 | - | 5 | 1027 | 8861322 | ||
| BC145236.1 | - | 4 | 863 | |||

Rnf135
Genomic Map