siRNA Details
| Gene Symbol: Zswim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028028.2 | NP_082304.1 | 2 | 733 | ||
| GenBank mRNA | AK010358.1 | - | 2 | 407 | ||
| AK149816.1 | - | 2 | 407 | |||
| AK150174.1 | - | 2 | 407 | |||
| BC046831.1 | - | 2 | 733 | 6812759 | ||

Zswim1
Genomic Map