siRNA Details
| Gene Symbol: Acad10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028037.4 | NP_082313.2 | 7 | 1095 | ||
| GenBank mRNA | AK050332.1 | - | 7 | 1299 | ||
| AK148195.1 | - | 7 | 1086 | |||
| BC027825.1 | - | 7 | 1071 | 5343012 | ||
| BC029047.1 | - | 3 | 244 | |||

Acad10
Genomic Map