siRNA Details
| Gene Symbol: Fbxo22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028049.2 | NP_082325.2 | 7 | 1309 | ||
| GenBank mRNA | AK002745.1 | - | 7 | 1189 | ||
| AK005473.1 | - | 6 | 1137 | |||
| AK211102.1 | - | 4 | 366 | |||
| AK213328.1 | - | 1 | 374 | |||
| BC018273.1 | - | 7 | 1249 | 4458296 | ||

Fbxo22
Genomic Map