siRNA Details
| Gene Symbol: Fndc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173182.2 | NP_775274.2 | 22 | 2869 | ||
| GenBank mRNA | AB098596.1 | - | 22 | 2807 | ||
| AK136013.1 | - | 1 | 67 | |||
| AK147346.1 | - | 23 | 2879 | |||
| AK220297.1 | - | 22 | 2869 | |||
| AY234860.1 | - | 4 | 611 | |||
| BC067389.1 | - | 22 | 2875 | 30431862 | ||
| DQ192037.1 | - | 21 | 2775 | |||

Fndc3b
Genomic Map