siRNA Details
| Gene Symbol: Phf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024250.4 | NP_077212.3 | 3, 4 | 378 | ||
| GenBank mRNA | AK007873.1 | - | 3, 4 | 358 | ||
| BF467016.1 | - | 3, 4 | 373 | |||
| CB247892.1 | - | 3, 4 | 378 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phf10| Gene Symbol: Phf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024250.4 | NP_077212.3 | 3, 4 | 378 | ||
| GenBank mRNA | AK007873.1 | - | 3, 4 | 358 | ||
| BF467016.1 | - | 3, 4 | 373 | |||
| CB247892.1 | - | 3, 4 | 378 | |||