siRNA Details
| Gene Symbol: Igsf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177886.1 | NP_001171357.1 | 3 | 229 | ||
| NM_001177887.1 | NP_001171358.1 | 3 | 233 | |||
| NM_028078.3 | NP_082354.1 | 3 | 226 | |||
| GenBank mRNA | AF537215.1 | - | 3 | 167 | ||
| AK008060.1 | - | 3 | 197 | |||
| AK078934.1 | - | 3 | 197 | |||
| AK086973.1 | - | 3 | 230 | |||
| BC004806.1 | - | 3 | 179 | 3584645 | ||
| BX511978.1 | - | 3 | 257 | |||
| BY033616.1 | - | 3 | 227 | |||

Igsf5
Genomic Map