siRNA Details
| Gene Symbol: 2010111I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289924.1 | NP_001276853.1 | 2 | 476 | ||
| NM_001289926.1 | NP_001276855.1 | 2 | 476 | |||
| GenBank mRNA | AJ810420.1 | - | 1 | 208 | ||
| AK044477.1 | - | 2 | 477 | |||
| AK141931.1 | - | 1 | 442 | |||
| BC141279.1 | - | 1 | 327 | 9055891 | ||

2010111I01Rik
Genomic Map