siRNA Details
| Gene Symbol: Dusp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028099.4 | NP_082375.4 | 6, 7 | 660 | ||
| GenBank mRNA | AK008492.1 | - | 1, 2 | 44 | ||
| AK081629.1 | - | 6, 7 | 637 | |||
| BC028640.1 | - | 2, 3 | 87 | |||
| BC067028.1 | - | 6, 7 | 614 | 30364073 | ||

Dusp11
Genomic Map