siRNA Details
| Gene Symbol: Ddhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028102.1 | NP_082378.1 | 10 | 1257 | ||
| GenBank mRNA | AK079311.1 | - | 9 | 1237 | ||
| AK173010.1 | - | 8 | 932 | |||
| BC046229.1 | - | 9 | 1177 | |||
| BC118962.2 | - | 10 | 1493 | 40095026 | ||

Ddhd2
Genomic Map