siRNA Details
| Gene Symbol: Dennd2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001093754.2 | NP_001087223.1 | 8 | 1046 | ||
| NM_001278941.1 | NP_001265870.1 | 9 | 1125 | |||
| NM_028110.2 | NP_082386.2 | 8 | 1044 | |||
| GenBank mRNA | AK008546.1 | - | 8 | 850 | ||
| AK050033.1 | - | 9 | 1126 | |||
| AK137891.1 | - | 1 | 1693 | |||
| AK157279.1 | - | 8 | 1047 | |||
| BC008266.1 | - | 8 | 1034 | 2811014 | ||
| BC043727.1 | - | 8 | 832 | 5035764 | ||

Dennd2d
Genomic Map