siRNA Details
Gene Symbol: Amer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028113.3 | NP_082389.1 | 1 | 933 | ||
GenBank mRNA | AK032651.1 | - | 1 | 874 | ||
AK139177.1 | - | 1 | 880 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Amer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028113.3 | NP_082389.1 | 1 | 933 | ||
GenBank mRNA | AK032651.1 | - | 1 | 874 | ||
AK139177.1 | - | 1 | 880 |