siRNA Details
Gene Symbol: Pex13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023651.4 | NP_076140.2 | 4 | 1217 | ||
GenBank mRNA | AF329877.2 | - | 4 | 1126 | ||
AJ304506.1 | - | 4 | 1150 | |||
AK011355.1 | - | 4 | 1187 | |||
AK029099.1 | - | 4 | 1187 | |||
AK029336.1 | - | 4 | 1190 | |||
AK031710.1 | - | 4 | 1217 | |||
AK031975.1 | - | 4 | 1173 | |||
AK032650.1 | - | 3 | 586 | |||
AK153438.1 | - | 4 | 1166 | |||
AK169148.1 | - | 4 | 1242 | |||
BC023683.1 | - | 4 | 1229 | 5343438 |
