siRNA Details
| Gene Symbol: Pygo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028116.2 | NP_082392.1 | 3 | 1033 | ||
| GenBank mRNA | AK011208.1 | - | 3 | 1035 | ||
| BC116630.1 | - | 3 | 870 | 40057035 | ||
| BC116631.1 | - | 3 | 870 | 40057038 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pygo1| Gene Symbol: Pygo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028116.2 | NP_082392.1 | 3 | 1033 | ||
| GenBank mRNA | AK011208.1 | - | 3 | 1035 | ||
| BC116630.1 | - | 3 | 870 | 40057035 | ||
| BC116631.1 | - | 3 | 870 | 40057038 | ||