siRNA Details
| Gene Symbol: Chst14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028117.3 | NP_082393.3 | 1 | 683 | ||
| GenBank mRNA | AK011230.1 | - | 1 | 620 | ||
| AK154555.1 | - | 1 | 672 | |||
| AK154636.1 | - | 1 | 652 | |||
| AK155115.1 | - | 2 | 591 | |||
| AK159357.1 | - | 1 | 651 | |||
| AK171614.1 | - | 1 | 553 | |||
| BC026886.1 | - | 1 | 504 | |||
| BC043700.1 | - | 1 | 656 | 5346204 | ||
| BC085479.1 | - | 1 | 600 | 6415106 | ||

Chst14
Genomic Map