siRNA Details
| Gene Symbol: Cep89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028120.2 | NP_082396.1 | 9, 10 | 1094 | ||
| GenBank mRNA | AK012067.1 | - | 9, 10 | 1095 | ||
| AK029107.1 | - | 9, 10 | 1125 | |||
| BC003981.1 | - | 9, 10 | 1149 | 3490246 | ||

Cep89
Genomic Map