siRNA Details
| Gene Symbol: Tbc1d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285991.1 | NP_001272920.1 | 19, 20 | 2235 | ||
| NM_001285993.1 | NP_001272922.1 | 18, 19 | 2136 | |||
| NM_028162.4 | NP_082438.3 | 18, 19 | 2169 | |||
| GenBank mRNA | AK154975.1 | - | 19, 20 | 2195 | ||
| AK163366.1 | - | 18, 19 | 2199 | |||
| AK220251.1 | - | 18, 19 | 2136 | |||
| BC043113.1 | - | 18, 19 | 1985 | 6407234 | ||
| BC098328.1 | - | 13, 14 | 1515 | 30357143 | ||

Tbc1d5
Genomic Map