siRNA Details
| Gene Symbol: Kcnk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029911.4 | NP_084187.2 | 3 | 525 | ||
| GenBank mRNA | AK006295.1 | - | 4 | 749 | ||
| AK019376.1 | - | 3 | 928 | |||
| AK031904.1 | - | 3 | 456 | |||
| AK036066.1 | - | 3 | 799 | |||
| AK082153.1 | - | 3 | 526 | |||
| BC132487.1 | - | 3 | 467 | 40130764 | ||
| BC137869.1 | - | 3 | 467 | 8860888 | ||
| DQ185134.1 | - | 3 | 422 | |||

Kcnk10
Genomic Map