siRNA Details
| Gene Symbol: Ccdc74a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166164.1 | NP_001159636.1 | 4 | 480 | ||
| GenBank mRNA | AK009347.1 | - | 4 | 333 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc74a| Gene Symbol: Ccdc74a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166164.1 | NP_001159636.1 | 4 | 480 | ||
| GenBank mRNA | AK009347.1 | - | 4 | 333 | ||