siRNA Details
| Gene Symbol: Amer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175179.4 | NP_780388.2 | 2 | 793 | ||
| GenBank mRNA | AK012651.1 | - | 2 | 778 | ||
| AK049774.1 | - | 2 | 778 | |||
| BC053442.1 | - | 2 | 764 | |||
| BC096616.1 | - | 1 | 539 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amer1| Gene Symbol: Amer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175179.4 | NP_780388.2 | 2 | 793 | ||
| GenBank mRNA | AK012651.1 | - | 2 | 778 | ||
| AK049774.1 | - | 2 | 778 | |||
| BC053442.1 | - | 2 | 764 | |||
| BC096616.1 | - | 1 | 539 | |||