siRNA Details
Gene Symbol: Zc2hc1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172414.4 | NP_766002.2 | 2 | 1480 | ||
GenBank mRNA | AK032455.1 | - | 2 | 1384 | ||
AK033213.1 | - | 2 | 1375 | |||
AK036602.1 | - | 2 | 1418 | |||
BC049940.1 | - | 2 | 1394 | 6305959 |
