siRNA Details
| Gene Symbol: 2210016L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028211.1 | NP_082487.1 | 4 | 314 | ||
| GenBank mRNA | AK008740.1 | - | 4 | 312 | ||
| AK135000.1 | - | 332 | ||||
| AK159360.1 | - | 4 | 312 | |||
| BC031162.1 | - | 4 | 314 | 4982352 | ||

2210016L21Rik
Genomic Map