siRNA Details
| Gene Symbol: 2210016L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028211.1 | NP_082487.1 | 6 | 565 | ||
| GenBank mRNA | AK135000.1 | - | 583 | |||
| AK159360.1 | - | 6 | 563 | |||
| BC031162.1 | - | 6 | 565 | 4982352 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210016L21Rik| Gene Symbol: 2210016L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028211.1 | NP_082487.1 | 6 | 565 | ||
| GenBank mRNA | AK135000.1 | - | 583 | |||
| AK159360.1 | - | 6 | 563 | |||
| BC031162.1 | - | 6 | 565 | 4982352 | ||