siRNA Details
| Gene Symbol: Wdr44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033628.1 | - | 2 | 225 | ||
| AK139877.1 | - | 7 | 875 | |||
| BC049191.1 | - | 16 | 2294 | 6476607 | ||
| BC068151.1 | - | 15 | 2490 | 30534873 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr44| Gene Symbol: Wdr44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK033628.1 | - | 2 | 225 | ||
| AK139877.1 | - | 7 | 875 | |||
| BC049191.1 | - | 16 | 2294 | 6476607 | ||
| BC068151.1 | - | 15 | 2490 | 30534873 | ||