siRNA Details
| Gene Symbol: Wdr44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC049191.1 | - | 9 | 1406 | 6476607 | |
| BC068151.1 | - | 8 | 1602 | 30534873 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr44| Gene Symbol: Wdr44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC049191.1 | - | 9 | 1406 | 6476607 | |
| BC068151.1 | - | 8 | 1602 | 30534873 | ||