siRNA Details
Gene Symbol: Lrpprc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028233.2 | NP_082509.2 | 22 | 2494 | ||
GenBank mRNA | AK013955.1 | - | 3 | 182 | ||
BC059862.1 | - | 22 | 2228 | 6417407 | ||
BC060199.1 | - | 22 | 2266 | |||
S81177.1 | - | 5 | 410 |
