siRNA Details
| Gene Symbol: Rab38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028238.7 | NP_082514.4 | 2 | 552 | ||
| GenBank mRNA | AB232634.1 | - | 2 | 423 | ||
| AK009296.1 | - | 2 | 513 | |||
| AK044783.1 | - | 2 | 515 | |||
| AK054253.1 | - | 2 | 553 | |||
| AK165682.1 | - | 2 | 555 | |||
| AK196794.1 | - | 1 | 183 | |||
| AK197116.1 | - | 1 | 183 | |||
| AY062237.1 | - | 2 | 512 | |||
| BC089578.1 | - | 2 | 512 | 6762755 | ||

Rab38
Genomic Map