siRNA Details
| Gene Symbol: Ssbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023672.2 | NP_076161.2 | 4 | 611 | ||
| NM_198438.1 | NP_940840.1 | 4 | 611 | |||
| GenBank mRNA | AF170906.1 | - | 4 | 341 | ||
| AF500117.1 | - | 4 | 341 | |||
| AK011853.1 | - | 4 | 581 | |||
| AK017711.1 | - | 4 | 612 | |||
| AK211107.1 | - | 2 | 61 | |||
| AY167987.1 | - | 4 | 224 | |||
| BC003430.1 | - | 4 | 365 | 2646149 | ||
| BC035475.1 | - | 4 | 554 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053358, XM_008763909, XM_008763905, XM_006238536, XM_008763906, and others.

Ssbp3
Genomic Map