siRNA Details
| Gene Symbol: Rnf219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026047.4 | NP_080323.3 | 6 | 1630 | ||
| GenBank mRNA | BC029231.1 | - | 6 | 1596 | 3156840 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf219| Gene Symbol: Rnf219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026047.4 | NP_080323.3 | 6 | 1630 | ||
| GenBank mRNA | BC029231.1 | - | 6 | 1596 | 3156840 | |