siRNA Details
Gene Symbol: 2610507B11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002004.2 | NP_001002004.2 | 18 | 3489 | ||
GenBank mRNA | AK122206.1 | - | 3 | 1141 | ||
BC040464.1 | - | 3 | 883 | |||
BC075633.1 | - | 18 | 3449 | 30537398 | ||
X81632.1 | - | 3 | 1398 |
