siRNA Details
| Gene Symbol: 2610507B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002004.2 | NP_001002004.2 | 7 | 794 | ||
| GenBank mRNA | AK149719.1 | - | 7 | 765 | ||
| AK150829.1 | - | 7 | 795 | |||
| BC019798.1 | - | 7 | 747 | |||
| BC075633.1 | - | 7 | 754 | 30537398 | ||
| BC080706.1 | - | 7 | 771 | |||

2610507B11Rik
Genomic Map